ENA183042478 Enamine

ethyl(2-ethylbutyl)amine

  • MDL number MFCD09943236
  • Cas Number : 854585-58-3
  • Molecular Weight : 129.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39880
Availability details for
Quantity
  • Availability
  • Add to Cart