ENA183042487 Enamine

3-{4-[1-(hydroxyimino)ethyl]phenyl}-1-phenylurea

  • Cas Number : 1016852-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39316
Availability details for
Quantity
  • Availability
  • Add to Cart