ENA183042496 Enamine

4-fluoro-3-sulfamoylbenzoic acid

  • MDL number MFCD01197510
  • Cas Number : 1535-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39319
Availability details for
Quantity
  • Availability
  • Add to Cart