ENA183157806 Enamine

N-[(oxolan-3-yl)methyl]-3,5-bis(trifluoromethyl)aniline

  • MDL number MFCD11505359
  • Cas Number : 1114822-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27397
Availability details for
Quantity
  • Availability
  • Add to Cart