ENA183158330 Enamine

2-Chloro-N-(3-trifluoromethyl-phenyl)-acetamide

  • MDL number MFCD00084932
  • Cas Number : 351-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61...63
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01496
Availability details for
Quantity
  • Availability
  • Add to Cart