ENA183888641 Enamine

3-amino-1-[4-(3-methylphenyl)piperazin-1-yl]propan-1-one hydrochloride

  • MDL number MFCD11505380
  • Cas Number : 1172301-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38843
Availability details for
Quantity
  • Availability
  • Add to Cart