ENA183888648 Enamine

ethyl 3-iodonaphthalene-2-carboxylate

  • MDL number MFCD11505489
  • Cas Number : 1071583-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39606
Availability details for
Quantity
  • Availability
  • Add to Cart