ENA183888652 Enamine

2-(7-bromo-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)acetic acid

  • MDL number MFCD11505526
  • Cas Number : 1110927-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39872
Availability details for
Quantity
  • Availability
  • Add to Cart