ENA183888654 Enamine

1-cyclohexyl-1-phenylmethanamine hydrochloride

  • MDL number MFCD11505530
  • Cas Number : 58271-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 330-331
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39890
Availability details for
Quantity
  • Availability
  • Add to Cart