ENA183888656 Enamine

3-(ethylamino)-2-methylpropanenitrile

  • MDL number MFCD11505532
  • Cas Number : 1099681-78-3
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39893
Availability details for
Quantity
  • Availability
  • Add to Cart