ENA183888657 Enamine

ethyl 1-(3-aminophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD11505533
  • Cas Number : 1153282-89-3
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39908
Availability details for
Quantity
  • Availability
  • Add to Cart