ENA183888665 Enamine

[1-(1,3-benzothiazol-2-yl)ethyl](methyl)amine hydrochloride

  • MDL number MFCD11505543
  • Cas Number : 1172922-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39955
Availability details for
Quantity
  • Availability
  • Add to Cart