ENA183888685 Enamine

4-[(3,5-dichloropyridin-2-yl)oxy]aniline hydrochloride

  • MDL number MFCD11505562
  • Cas Number : 1171554-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-40205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40205
Availability details for
Quantity
  • Availability
  • Add to Cart