ENA183888686 Enamine

(2-methylpropyl)(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD11505564
  • Cas Number : 1177279-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40217
Availability details for
Quantity
  • Availability
  • Add to Cart