ENA183888687 Enamine

N-(4-fluorophenyl)-N'-piperidin-4-ylurea hydrochloride

  • MDL number MFCD11505565
  • Cas Number : 913634-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227...229
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40219

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40219
Availability details for
Quantity
  • Availability
  • Add to Cart