ENA183888689 Enamine

N-[4-(1-aminoethyl)phenyl]cyclopropanecarboxamide hydrochloride

  • MDL number MFCD11505567
  • Cas Number : 1170962-96-5
  • Molecular Weight : 240.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40230
Availability details for
Quantity
  • Availability
  • Add to Cart