ENA183888691 Enamine

2-(3,4-dimethoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid hydrochloride

  • MDL number MFCD11505570
  • Cas Number : 1170288-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-40255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40255
Availability details for
Quantity
  • Availability
  • Add to Cart