ENA183888697 Enamine

2-{[(2-chloro-6-fluorophenyl)methyl]amino}propanoic acid hydrochloride

  • MDL number MFCD11505576
  • Cas Number : 1396966-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40285
Availability details for
Quantity
  • Availability
  • Add to Cart