ENA183888699 Enamine

3,4-dimethyl 2-(4-aminophenyl)-1-methyl-1H-pyrrole-3,4-dicarboxylate

  • MDL number MFCD11505578
  • Cas Number : 1110865-98-9
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40294

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-40294
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart