ENA183888700 Enamine

3-(aminomethyl)-N-phenylbenzamide hydrochloride

  • MDL number MFCD11505579
  • Cas Number : 1171573-70-8
  • Molecular Weight : 262.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40295
Availability details for
Quantity
  • Availability
  • Add to Cart