ENA183888709 Enamine

6,7-dimethyl 5-(3-aminophenyl)-2,3-dihydro-1H-pyrrolizine-6,7-dicarboxylate

  • MDL number MFCD11505588
  • Cas Number : 1110927-76-8
  • Molecular Weight : 314.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40372
Availability details for
Quantity
  • Availability
  • Add to Cart