ENA183888718 Enamine

2-(1-aminocyclobutyl)-1H-imidazole-4-carboxylic acid

  • MDL number MFCD11505598
  • Cas Number : 1110909-14-2
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40407
Availability details for
Quantity
  • Availability
  • Add to Cart