ENA183888724 Enamine

2-amino-N-(3-fluoro-4-methylphenyl)ethanesulfonamide hydrochloride

  • MDL number MFCD11505606
  • Cas Number : 1171345-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40457

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40457
Availability details for
Quantity
  • Availability
  • Add to Cart