ENA183888726 Enamine

2-amino-N,N,4-trimethylpentanamide hydrochloride

  • MDL number MFCD11505609
  • Cas Number : 1214130-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40489
Availability details for
Quantity
  • Availability
  • Add to Cart