ENA183888729 Enamine

4-[4-(2-methylpropyl)benzoyl]piperidine hydrochloride

  • MDL number MFCD11505612
  • Cas Number : 1171984-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40494
Availability details for
Quantity
  • Availability
  • Add to Cart