ENA183888732 Enamine

4-(piperidine-4-carbonyl)phenol hydrochloride

  • MDL number MFCD11505615
  • Cas Number : 1171519-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40502
Availability details for
Quantity
  • Availability
  • Add to Cart