ENA183888740 Enamine

4-bromo-1-(2-fluorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11505624
  • Cas Number : 1157114-86-7
  • Molecular Weight : 285.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40810
Availability details for
Quantity
  • Availability
  • Add to Cart