ENA183888747 Enamine

2-({4-methyl-5,12-dithia-3-azatricyclo[7.3.0.0,2,6]dodeca-1,3,6,8,10-pentaen-8-yl}oxy)ethan-1-amine dihydrochloride

  • MDL number MFCD11505630
  • Cas Number : 1171669-99-0
  • Molecular Weight : 337.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40854
Availability details for
Quantity
  • Availability
  • Add to Cart