ENA183888748 Enamine

2-(2,6-dimethoxyphenoxy)ethan-1-amine hydrochloride

  • MDL number MFCD11505631
  • Cas Number : 87780-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40856
Availability details for
Quantity
  • Availability
  • Add to Cart