ENA183888751 Enamine

methyl 3-[(chlorosulfonyl)methyl]-2-furoate

  • MDL number MFCD11505634
  • Cas Number : 1114823-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40928

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40928
Availability details for
Quantity
  • Availability
  • Add to Cart