ENA183888764 Enamine

N-(2-bromo-4,6-difluorophenyl)-2-chloroacetamide

  • MDL number MFCD04127864
  • Cas Number : 737780-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24571

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24571
Availability details for
Quantity
  • Availability
  • Add to Cart