ENA183888773 Enamine

N-[2-(1H-1,3-benzodiazol-2-yl)ethyl]-2-methylbenzamide

  • MDL number MFCD01034014
  • Cas Number : 303991-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38858
Availability details for
Quantity
  • Availability
  • Add to Cart