ENA183888778 Enamine

N-[3-(1H-1,3-benzodiazol-2-yl)propyl]butanamide

  • MDL number MFCD04459338
  • Cas Number : 686736-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38865
Availability details for
Quantity
  • Availability
  • Add to Cart