ENA183888783 Enamine

3-(1H-1,3-benzodiazol-2-yl)-3-hydroxypropanoic acid

  • MDL number MFCD00458808
  • Cas Number : 99072-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-38870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38870
Availability details for
Quantity
  • Availability
  • Add to Cart