ENA183888795 Enamine

bis(propan-2-yl) [(4-methylbenzenesulfonyl)oxy]methanephosphonate

  • MDL number MFCD06660915
  • Cas Number : 35717-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-32
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-39109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39109
Availability details for
Quantity
  • Availability
  • Add to Cart