ENA183888800 Enamine

1-(piperidine-4-carbonyl)piperidine-4-carboxamide

  • MDL number MFCD06740222
  • Cas Number : 886508-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39231
Availability details for
Quantity
  • Availability
  • Add to Cart