ENA183888803 Enamine

1-(4-aminophenyl)imidazolidin-2-one

  • MDL number MFCD09809796
  • Cas Number : 89518-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25880
Availability details for
Quantity
  • Availability
  • Add to Cart