ENA183888811 Enamine

4-(1,3-benzoxazol-2-yl)butanoic acid

  • MDL number MFCD02656550
  • Cas Number : 288569-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-39457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39457
Availability details for
Quantity
  • Availability
  • Add to Cart