ENA183888818 Enamine

1-(2-hydroxyethyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11121120
  • Cas Number : 1152571-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39632
Availability details for
Quantity
  • Availability
  • Add to Cart