ENA183888834 Enamine

4-[(3-bromobenzoyl)amino]-2-hydroxybenzoic acid

  • MDL number MFCD09945824
  • Cas Number : 1016804-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39822
Availability details for
Quantity
  • Availability
  • Add to Cart