ENA183888835 Enamine

4-(2-amino-1,3-thiazol-4-yl)benzamide

  • MDL number MFCD11054234
  • Cas Number : 1065101-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39825
Availability details for
Quantity
  • Availability
  • Add to Cart