ENA183888843 Enamine

2-(1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD03086184
  • Cas Number : 55317-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39877

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39877
Availability details for
Quantity
  • Availability
  • Add to Cart