ENA183888845 Enamine

3,3-dimethyl-1-phenylbutan-1-amine

  • MDL number MFCD09809111
  • Cas Number : 727964-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39888
Availability details for
Quantity
  • Availability
  • Add to Cart