ENA183888848 Enamine

2-chloro-N-(2-methyl-1,3-dioxo-2,3-dihydro-1H-isoindol-5-yl)acetamide

  • MDL number MFCD04133251
  • Cas Number : 828299-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22670
Availability details for
Quantity
  • Availability
  • Add to Cart