ENA183888850 Enamine

4-methoxynaphthalene-1-sulfonyl chloride

  • MDL number MFCD08752807
  • Cas Number : 56875-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39897
Availability details for
Quantity
  • Availability
  • Add to Cart