ENA183888859 Enamine

1-(cyclohexylmethyl)piperidin-4-one

  • MDL number MFCD09947739
  • Cas Number : 64306-76-9
  • Molecular Weight : 195.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39911
Availability details for
Quantity
  • Availability
  • Add to Cart