ENA183888869 Enamine

methyl 3-fluoroimidazo[1,2-a]pyridine-2-carboxylate

  • MDL number MFCD03856166
  • Cas Number : 695212-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39925
Availability details for
Quantity
  • Availability
  • Add to Cart