ENA183888880 Enamine

[(thien-2-ylmethyl)sulfonyl]acetic acid

  • MDL number MFCD08444899
  • Cas Number : 180044-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39949
Availability details for
Quantity
  • Availability
  • Add to Cart