ENA183888893 Enamine

2-[(2-methylpropyl)sulfanyl]acetic acid

  • MDL number MFCD08445180
  • Cas Number : 20600-62-8
  • Molecular Weight : 148.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39969
Availability details for
Quantity
  • Availability
  • Add to Cart