ENA183888895 Enamine

4-(6-bromo-4-oxo-3,4-dihydroquinazolin-2-yl)butanoic acid

  • MDL number MFCD09042596
  • Cas Number : 926193-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39972
Availability details for
Quantity
  • Availability
  • Add to Cart